Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523983
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523983 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523983 Supplier Invitrogen™ Supplier No. PA523983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E2F2 Polyclonal Antibody for Western Blot

E2F2 is a member of the E2Ffamily of transcription factors. The E2F family plays a crucialrole in the control of cell cycle and action of tumor suppressorproteins and is also a target of the transforming proteins of smallDNA tumor viruses. The E2F proteins contain several evolutionallyconserved domains found in most members of the family. Thesedomains include a DNA binding domain, a dimerization domain whichdetermines interaction with the differentiation regulatedtranscription factor proteins (DP), a transactivation domainenriched in acidic amino acids, and a tumor suppressor proteinassociation domain which is embedded within the transactivationdomain. This protein and another 2 members, E2F1 and E2F3, have anadditional cyclin binding domain. This protein binds specificallyto retinoblastoma protein pRB in a cell-cycle dependent manner, andit exhibits overall 46% amino acid identity to E2F1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene E2F2
Gene Accession No. P56931, Q14209
Gene Alias E2F transcription factor 2; E2F2; E2F-2; LOW QUALITY PROTEIN: transcription factor E2F2; transcription factor E2F2
Gene Symbols E2F2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 258-286 amino acids from the central region of human E2F2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1870, 242705
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.