Learn More
Invitrogen™ E2F2 Recombinant Rabbit Monoclonal Antibody (1H7D6)

Description
Positive Samples: HeLa, SH-SY5Y, Mouse lung, Mouse thymus.
Specifications
Specifications
| Antigen | E2F2 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1H7D6 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | E2F2 |
| Gene Accession No. | P56931, Q14209 |
| Gene Alias | E2F transcription factor 2; E2F2; E2F-2; LOW QUALITY PROTEIN: transcription factor E2F2; transcription factor E2F2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.