Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F4/E2F5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536051
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536051 Supplier Invitrogen™ Supplier No. PA536051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 44 kDa. Purity is >95% by SDS-PAGE.

E2F was originally identified through its role in transcriptional activation of the adenovirus E2 promoter. Sequences homologous to the E2F binding site have been found upstream of a number of genes that encode proteins with putative functions in the G1 and S phases of the cell cycle. E2F-1 is a member of a broader family of transcriptional regulators including E2F-2, E2F-3, E2F-4, E2F-5 and E2F-6, each of which forms heterodimers with a second protein, DP-1, forming an active E2F transcriptional regulatory complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F4/E2F5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene E2F4
Gene Accession No. Q15329, Q16254, Q61502, Q62814, Q8R0K9
Gene Alias 2010111M04Rik; AI427446; AU024671; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2F4; E2F-4; E2f5; E2F-5; p107/p130-binding protein; transcription factor E2F4; Transcription factor E2F5
Gene Symbols E2F4, E2F5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 18-72 of human E2F4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360427, 104394, 116651, 13559, 1874, 1875
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.