Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F4 Monoclonal Antibody (4E2F04 (WUF10)), Invitrogen™

Catalog No. PIMA511276
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511276 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511276 Supplier Thermo Scientific Supplier No. MA511276

Mouse Monoclonal Antibody

MA5-11276 targets E2F-4 Transcription Factor in GS, IF, IHC (P), and WB applications and shows reactivity with Human and Rat samples. The MA5-11276 immunogen is recombinant human E2F-4 protein.

E2F4 Monoclonal antibody specifically detects E2F4 in Human, Mouse, Rat samples. It is validated for Gel Shift, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F4
Applications Gel Shift, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4E2F04 (WUF10)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene E2f4
Gene Accession No. Q16254, Q8R0K9
Gene Alias 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4
Gene Symbols E2f4
Host Species Mouse
Immunogen Recombinant human E2F-4 protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360427, 104394, 1874
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.