Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E2F4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579181
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Lung Tissue, SMMC whole cell.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
E2F4 | |
Polyclonal | |
Unconjugated | |
E2f4 | |
2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100360427, 1874 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5 mg BSA and 0.05 mg sodium azide, 0.05 mg thimerosal | |
Q16254 | |
E2f4 | |
A synthetic peptide corresponding to a sequence in the middle region of human E2F4 (228-243aa LPKPALAQSQEASRPN). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction