Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F4 Recombinant Rabbit Monoclonal Antibody (24E12)
GREENER_CHOICE

Catalog No. FEMA550001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550001 Supplier Invitrogen™ Supplier No. MA550001
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

E2F4 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24E12
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene E2f4
Gene Accession No. Q16254
Gene Alias 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4
Gene Symbols E2f4
Host Species Rabbit
Immunogen A synthesized peptide derived from human E2F4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1874
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.