Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F4 Recombinant Rabbit Monoclonal Antibody (3O6Y9)
E2F4 Recombinant Rabbit Monoclonal Antibody (3O6Y9)
Supplier: Thermo Scientific MA542723
This item is not returnable.
View return policy
Description
E2F4 Recombinant Monoclonal Antibody for Western Blot

Specifications
E2F4 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16254, Q8R0K9 | |
E2f4 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human E2F4 (Q16254). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
3O6Y9 | |
Unconjugated | |
E2f4 | |
2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
104394, 1874 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction