Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F5 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299198AF488
Description
E2F5 Polyclonal specifically detects E2F5 in Human samples. It is validated for Western Blot.Specifications
E2F5 | |
Polyclonal | |
Western Blot | |
E2F transcription factor 5, p130-binding, E2F-5, transcription factor E2F5 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human E2F5. | |
0.1 mL | |
Cell Cycle and Replication | |
1875 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction