Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F5 Polyclonal Antibody, Abbomax

Catalog No. 502203803
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3803 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3803 Supplier Abbomax Inc Supplier No. 5007534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Breast Carcinoma Cellular location: Nuclear.

E2F5 Polyclonal antibody specifically detects E2F5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen E2F5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene E2F5
Gene Accession No. Q15329, Q61502, Q62814
Gene Alias AU024671; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2f5; E2F-5; Transcription factor E2F5
Host Species Rabbit
Immunogen A synthetic peptide derived from internal segment of human E2F5
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116651, 13559, 1875
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.