Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA581166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581166 Supplier Thermo Scientific Supplier No. PA581166

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human E2F5.

E2F5 Polyclonal antibody specifically detects E2F5 in Human samples. It is validated for Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene E2F5
Gene Accession No. Q15329
Gene Alias AU024671; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2f5; E2F-5; Transcription factor E2F5
Gene Symbols E2F5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human E2F5
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1875
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.