Learn More
E2F5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA132480
Description
PA1-32480 detects E2F5 from human samples. PA1-32480 has been successfully used in Western blot applications. The PA1-32480 immunogen is: Synthetic peptide derived from residues 126-180 of Human E2F5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionarily conserved domains that are present in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein is differentially phosphorylated and is expressed in a wide variety of human tissues. It has higher identity to E2F4 than to other family members. Both this protein and E2F4 interact with tumor suppressor proteins p130 and p107, but not with pRB. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
E2F5 | |
Polyclonal | |
Unconjugated | |
Q15329 | |
E2F5 | |
Synthetic peptide derived from residues 126-180 of Human E2F5. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
E2F5 | |
E2F-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.