Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F6 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299406APC
Description
E2F6 Polyclonal antibody specifically detects E2F6 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
E2F6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human E2F6 | |
0.1 mL | |
Cell Cycle and Replication | |
1876 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction