Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F6 Antibody (PCRP-E2F6-1F8), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308952MFV450
Description
E2F6 Monoclonal antibody specifically detects E2F6 in Human samples. It is validated for Flow CytometrySpecifications
E2F6 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-E2F6-1F8 | |
Flow Cytometry | |
E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
Recombinant full-length human E2F6 protein (Uniprot: O75461) | |
0.1 mL | |
Cell Cycle and Replication | |
1876 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction