Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F6 Antibody (SR1531), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321950100UL
Description
E2F6 Monoclonal antibody specifically detects E2F6 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
E2F6 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1531 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
A synthesized peptide derived from human E2F6 (Uniprot #: O75461) | |
100 μg | |
Cell Cycle and Replication | |
1876 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction