Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F6 Mouse anti-Human, Clone: PCRP-E2F6-1F8, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | E2F6 |
|---|---|
| Clone | PCRP-E2F6-1F8 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
E2F6 Monoclonal specifically detects E2F6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| E2F6 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 1876 | |
| IgG2b κ | |
| Protein A or G purified |
| PCRP-E2F6-1F8 | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
| Recombinant full-length human E2F6 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title