Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540507 Supplier Invitrogen™ Supplier No. PA540507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HEQIVIAVKA PAETRLDVPA PREDSITVHI RSTNGPIDVY LCEVEQGQTS Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

This gene encodes a member of the E2F transcription factor protein family. E2F family members play a crucial role in control of the cell cycle and of the action of tumor suppressor proteins. They are also a target of the transforming proteins of small DNA tumor viruses. Many E2F proteins contain several evolutionarily conserved domains: a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The encoded protein of this gene is atypical because it lacks the transactivation and tumor suppressor protein association domains. It contains a modular suppression domain and is an inhibitor of E2F-dependent transcription. The protein is part of a multimeric protein complex that contains a histone methyltransferase and the transcription factors Mga and Max. Multiple transcript variants have been reported for this gene, but it has not been clearly demonstrated that they encode valid isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene E2F6
Gene Accession No. O75461
Gene Alias AI462434; E2F transcription factor 6; E2f6; E2F-6; E2F6b; E2F-binding site-modulating activity protein; EMA; MGC111545; Transcription factor E2F6; transcriptional repressor E2F6
Gene Symbols E2F6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human E2F6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1876
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.