Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F7 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152648H
Description
E2F7 Polyclonal antibody specifically detects E2F7 in Human samples. It is validated for Western BlotSpecifications
E2F7 | |
Polyclonal | |
Western Blot | |
E2F transcription factor 7, E2F-7, FLJ12981, transcription factor E2F7 | |
The immunogen for this product maps to a region between residue 1 and 50 of human E2F Transcription Factor 7 using the numbering given in entry NP_976328.2 (GeneID 144455). | |
0.1 mL | |
Signal Transduction | |
144455 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction