Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F7 Polyclonal Antibody
E2F7 Polyclonal Antibody
Supplier: Thermo Scientific PA550495
Description
E2F7 Polyclonal Antibody for IHC (P)

Specifications
E2F7 | |
Polyclonal | |
Unconjugated | |
E2F7 | |
A630014C11Rik; D10Ertd739e; E2F transcription factor 7; E2f7; E2F-7; fc43h03; FLJ12981; no; si:dkey-217k21.2; Transcription factor E2F7; wu:fc43h03 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144455 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96AV8 | |
E2F7 | |
Synthetic peptide corresponding to a region derived from internal residues of human E2F transcription factor 7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction