Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568911 Supplier Invitrogen™ Supplier No. PA568911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Horse: 85%; Human: 86%; Mouse: 100%; Pig: 92%; Rabbit: 79%; Rat: 86%.

E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene E2F7
Gene Accession No. Q6S7F2, Q96AV8
Gene Alias A630014C11Rik; D10Ertd739e; E2F transcription factor 7; E2f7; E2F-7; fc43h03; FLJ12981; no; si:dkey-217k21.2; Transcription factor E2F7; wu:fc43h03
Gene Symbols E2F7
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse E2F7.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144455, 52679
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.