Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E2F7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114731
Description
Antibody detects endogenous levels of total E2F7.
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.
Specifications
E2F7 | |
Polyclonal | |
Unconjugated | |
E2F7 | |
A630014C11Rik; D10Ertd739e; E2F transcription factor 7; E2f7; E2F-7; fc43h03; FLJ12981; no; si:dkey-217k21.2; Transcription factor E2F7; wu:fc43h03 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
144455, 314818, 52679 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
D4A4D7, Q6S7F2, Q96AV8 | |
E2F7 | |
A synthesized peptide derived from human E2F7(Accession Q96AV8), corresponding to amino acid residues N33-P83. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction