Learn More
E2F7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303037A

Description
The recommended shelf life for this product is 1 year from date of receipt.
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression .Specifications
E2F7 | |
Polyclonal | |
Unconjugated | |
E2F7 | |
E2F7; E2F transcription factor 7; FLJ12981 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144455 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96AV8 | |
E2F7 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.