Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F8 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152650AF750
Description
E2F8 Polyclonal antibody specifically detects E2F8 in Human samples. It is validated for Western BlotSpecifications
E2F8 | |
Polyclonal | |
Western Blot | |
E2F family member 8, E2F transcription factor 8, E2F-8, FLJ23311, transcription factor E2F8 | |
The immunogen for this product maps to a region between residue 817 and 867 of human E2F Transcription Factor 8 using the numbering given in entry NP_078956.2 (GeneID 79733). | |
0.1 mL | |
Cell Cycle and Replication | |
79733 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction