Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E2F8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588117 Supplier Invitrogen™ Supplier No. PA588117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVPGQSQPNG QSVAVTGAQQ PVPVTPKGSQ LVAESFFRTP GGPTKPTSSS CMDFEGANKT SLGTLFVPQR KLEVSTEDVH Positive Samples: SW480, Mouse testis; Cellular Location: Nucleus.

E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation (Christensen et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene E2F8
Gene Accession No. A0AVK6, Q58FA4
Gene Alias 4432406C08Rik; AA410048; E2F 8; E2F family member 8; E2F transcription factor 8; E2F8; E2F-8; FLJ23311; RGD1308091; Transcription factor E2F8
Gene Symbols E2F8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 788-867 of human E2F8 (NP_0789562).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108961, 79733
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.