Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E2F8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100878
Description
Antibody detects endogenous levels of total E2F8.
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation (Christensen et al., 2005).
Specifications
E2F8 | |
Polyclonal | |
Unconjugated | |
E2F8 | |
4432406C08Rik; AA410048; E2F 8; E2F family member 8; E2F transcription factor 8; E2F8; E2F-8; FLJ23311; RGD1308091; Transcription factor E2F8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79733 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A0AVK6 | |
E2F8 | |
A synthesized peptide derived from human E2F8(Accession A0AVK6), corresponding to amino acid residues Q807-R857. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction