Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ E3 Ubiquitin Ligase BRE1B, Mouse, Unlabeled, Clone: 4H8.1,

Catalog No. MABE949MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE949MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE949MI Supplier MilliporeSigma™ Supplier No. MABE949
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E3 ubiquitin-protein ligase BRE1B (UniProt O75150; also known as 95kDa retinoblastoma-associated protein, BRE1-B, RBP95, RING finger protein 40, Syntaxin-1-interacting RING finger protein, Protein staring) is encoded by the RNF40 (also known as BRE1B, KIAA0661) gene (Gene ID 9810) in human. Proteini ubiquitination is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type (really interesting new gene), HECT-type (homologous to E6-AP C-terminus), and U-box type of E3 ligases. RING finger ubiquitin ligase members can function as monomers, dimers, or multi-subunit complexes. The RNF20-RNF40 heterodimer mediates the ubiquitination of histone H2B at Lysine 120, an event that plays an important role in decompacting chromatin fiber to allow transcription elongation, in stem cell differentiation, as well as in DNA double-strand breaks (DSBs) repair via the homologous recombination (HR) and nonhomologous end-joining (NHEJ) pathways. Human RNF40 is a 1001-amino acid protein containing a RING-type zinc finger in its C-terminal region (a.a. 948-987) that mediates its interaction with RNF20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E3 Ubiquitin Ligase BRE1B
Applications Western Blot
Classification Monoclonal
Clone 4H8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RNF40, BRE1B, KIAA0661
Gene Accession No. O75150
Host Species Mouse
Immunogen GST-tagged recombinant internal fragment from the C-terminal half of human BRE1B .
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001193962
Test Specificity Clone 4H8.1 epitope region is present in all three spliced isoforms of human BRE1B reported by UniProt (O75150).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.