Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E3 ubiquitin-protein ligase RNF31, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABS148625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS148625UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS148625UG Supplier MilliporeSigma Supplier No. ABS148625UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-E3 ubiquitin-protein ligase RNF31, Cat. No. ABS1486, is a rabbit polyclonal antibody that detects E3 ubiquitin-protein ligase RNF31 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

E3 ubiquitin-protein ligase RNF31 (UniProt: Q96EP0; also known as EC: 2.3.2.27, HOIL-1-interacting protein, HOIP, RING finger protein 31, RING-type E3 ubiquitin transferase RNF31, Zinc in-between-RING-finger ubiquitin-associated domain protein) is encoded by the RNF31 (also known as ZIBRA) gene (Gene ID: 55072) in human. E3 ubiquitin-protein ligase RNF31 is a cytoplasmic protein that is a component of the linear ubiquitin chain assembly complex (LUBAC complex), which conjugates linear ('Met-1'-linked) polyubiquitin chains to substrates and plays a key role in NF-kappa-B activation and regulation of inflammation. LUBAC conjugates linear polyubiquitin to IKBKG (NEMO) and receptor interacting serine/threonine Kinase 1 (RIPK1) and is involved in activation of the canonical NF-kappa-B and the JNK signaling pathways. LUBAC-mediated polyubiquitination interferes with TNF-induced cell death and thereby prevents inflammation. Together with OUTLIN deubiquitinase, the LUBAC complex is also reported to regulate the canonical Wnt signaling pathway during angiogenesis. E3 ubiquitin-protein ligase RNF31 is expressed in both normal and transformed breast epithelial cell lines. It contains a polyubiquitin-binding (PUB) domain (aa 71-142) that mediates interaction with the PCNA-interacting motifs of C-terminus of the AAA+ ATPase p97 (VCP) and RNF31, with a strong preference for RNF3. It also has a ubiquitin-associated (UBA) domain (aa 564-615) that mediates association with RanBP-type and C3HC4-type zinc finger-containing protein 1 (RBCK1/HOIL1) via interaction with its ubiquitin-like (UBL) domain. (Ref.: Elliot, PR et al (2014). Mol. Cell 54(3); 335-48).
TRUSTED_SUSTAINABILITY

Specifications

Antigen E3 ubiquitin-protein ligase RNF31
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected E3 ubiquitin-protein ligase RNF31 in human adrenal gland and human uterus tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias EC: 2.3.2.27;HOIL-1-interacting protein;HOIP;RING finger protein 31;RING-type E3 ubiquitin transferase RNF31;Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Symbols RNF31;ZIBRA
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human E3 ubiquitin-protein ligase RNF31.
Molecular Weight of Antigen ∽120 kDa observed; 119.65 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase RNF31 in human cells. It targets an epitope within 17 amino acids from the C-terminal region. Immunogen sequence is conserved in all 3 isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.