Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E3 ubiquitin-protein ligase UBR1 Mouse anti-Human, Rat, Clone: 6H9.1, MilliporeSigma™

Catalog No. MABS118025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1180-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1180-25 Supplier MilliporeSigma Supplier No. MABS118025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-E3 ubiquitin-protein ligase UBR1, clone 6H9.1 is a mouse monoclonal that detects E3 ubiquitin-protein ligase UBR1 and has been tested for use in Immunohistochemistry (Paraffin) Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E3 ubiquitin-protein ligase UBR1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H9.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols UBR1
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 101 amino acids from the internal region of human E3 ubiquitin-protein ligase UBR1.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_777576
Test Specificity Clone 6H9.1 detects E3 ubiquitin-protein ligase UBR1 in human and rat cells. It targets an epitope within 101 amino acids from the internal region.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.