Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144504 Supplier Invitrogen™ Supplier No. PA5144504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E3L Polyclonal Antibody for ELISA, Western Blot

RNA-binding protein that plays a role in the inhibition of multiple cellular antiviral responses activated by double-stranded RNA (dsRNA), such as inhibition of PKR activation, necroptosis, and IFN-mediated antiviral activities. Recognizes and binds Z-RNA structures via its Z-binding domain and dsRNA via its DRBM domain: RNA-binding activity is required to escape host ZBP1-dependent necroptosis. Mechanistically, the Z-binding domain binds Z-RNAs that are produced during vaccinia virus infection, thereby competing with Z-RNA detection by host ZBP1, suppressing ZBP1-dependent necroptosis. Acts as a key inhibitor of the interferon response by blocking the phosphorylation and subsequent activation of IRF3 and IRF7 kinases that are required for interferon-alpha gene expression. Inhibits NF-kappa-B activation and the ubiquitin-like protein ISG15, which is an early antiviral protein. The binding with host ISG15 subsequently blocks host ISGylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E3L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Protein E3
Host Species Rabbit
Immunogen Recombinant Vaccinia virus Protein E3 (E3L) (1-190AA).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.