Learn More
E4BP4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302606AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Expression of interleukin-3 is restricted to activated T cells, natural killer cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site .Specifications
E4BP4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q16649 | |
NFIL3 | |
Between 412 and 462 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NFIL3 | |
E4BP4, IL3BP1, NF-IL3A, NFIL3A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4783 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.