Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E4F1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002871AF750
Description
E4F1 Polyclonal antibody specifically detects E4F1 in Human samples. It is validated for Western BlotSpecifications
E4F1 | |
Polyclonal | |
Western Blot | |
E4F transcription factor 1p50E4F, E4FMGC99614, EC 6.3.2.-, p120E4F, Putative E3 ubiquitin-protein ligase E4F1, Transcription factor E4F, transcription factor E4F1 | |
The immunogen recognized by this antibody maps to a region between residue 725 and the C-terminus (residue 784) of human E4F1 (p120E4F) using the numbering given in entry NP_004415.1 (GeneID 1877). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1877 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction