Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E4F1 Antibody (PCRP-E4F1-2D1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308349MFV610
Description
E4F1 Monoclonal antibody specifically detects E4F1 in Human samples. It is validated for Flow CytometrySpecifications
E4F1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
1877 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
PCRP-E4F1-2D1 | |
Flow Cytometry | |
E4F transcription factor 1p50E4F, E4FMGC99614, EC 6.3.2.-, p120E4F, Putative E3 ubiquitin-protein ligase E4F1, Transcription factor E4F, transcription factor E4F1 | |
Recombinant full-length human E4F1 protein (Uniprot: Q66K89) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction