Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E4F1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567167
Description
Immunogen sequence: RHLKSLTPCTE KIRFSVSKDV VVSKEDARAG SGAGAAGLGT ATSSVTGEPI ETSPVIHLVT DAKGTVIHEV HVQM Highest antigen sequence identity to the following orthologs - mouse 77%, rat 79%.
May function as a transcriptional repressor. May also function as a ubiquitin ligase mediating ubiquitination of chromatin-associated TP53. Functions in cell survival and proliferation through control of the cell cycle. Functions in the p53 and pRB tumor suppressor pathways and regulates the cyclin CCNA2 transcription.; FUNCTION: Identified as a cellular target of the adenoviral oncoprotein E1A, it is required for both transcriptional activation and repression of viral genes.
Specifications
E4F1 | |
Polyclonal | |
Unconjugated | |
E4F1 | |
deoxyribonuclease 1-like 2; Dnase1l2; E4F; E4F transcription factor 1; E4F1; p120E4F; p50E4F; phi-AP3; putative E3 ubiquitin-protein ligase E4F1; RING-type E3 ubiquitin transferase E4F1; Transcription factor E4F; transcription factor E4F1; Transcription factor phi AP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1877 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q66K89 | |
E4F1 | |
Recombinant Human E4F1. Recombinant protein control fragment (Product #RP-105531). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction