Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E6AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595918 Supplier Invitrogen™ Supplier No. PA595918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLIRLWSKYN ADQIRRMMET FQQLITYKVI SNEFNSRNLV NDDDAIVAAS KCLKMVYYAN VVGGEVDTNH NEEDDEEPIP ESSELTLQEL LGEERRNKKG PRVDPLETEL GVKTLDCRKP LIPFEEFINE PLNEVLEMDK DYTFFKVETE NKFSFMTCPF ILNAVTKNLG LYYDNRIRMY SERRITVLYS LVQGQQLNPY LRLKVRRDHI IDDALVRLEM IAMENPADLK KQLYVEFEGE QGVDEGGVSK EFFQLVVEEI FNPDIGMFTY DESTKLFWFN PSSFETEGQF T; Positive Samples: 293T, SKOV3, BT-474, U-251MG, Jurkat, Mouse brain, Mouse heart, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus.

UBE3A (ubiquitin-protein ligase E3A, AKA E6AP) is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and transfers it to its substrates. Several substrates have been identified including the ARNTL/BMAL1, ARC, RAD23A, RAD23B, MCM7, annexin A1, the PML tumor suppressor, and the cell cycle regulator CDKN1B. Additionally, UBE3A may function as a cellular quality control ubiquitin ligase by helping the degradation of the cytoplasmic misfolded proteins. UBE3A also plays an important role in the regulation of the circadian clock. Defects in UBE3A are a cause of Angelman syndrome (AS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen E6AP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE3A
Gene Accession No. O08759, Q05086
Gene Alias 4732496B02; 5830462N02Rik; A130086L21Rik; ANCR; AS; CTCL tumor antigen se37-2; E6AP; E6-AP; E6-AP ubiquitin protein ligase; E6AP ubiquitin-protein ligase; EPVE6AP; FLJ26981; HECT-type ubiquitin transferase E3A; HPVE6A; human papilloma virus E6-associated protein; human papillomavirus E6-associated protein; mKIAA4216; oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; UBE3A; ubiquitin conjugating enzyme E3A; ubiquitin protein ligase E3A; ubiquitin-protein ligase E3A
Gene Symbols UBE3A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-590 of human UBE3A (NP_5708531).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22215, 7337
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.