Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E6AP/UBE3A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500239G
Description
E6AP/UBE3A Polyclonal antibody specifically detects E6AP/UBE3A in Human, Mouse samples. It is validated for Western BlotSpecifications
E6AP/UBE3A | |
Polyclonal | |
Western Blot | |
ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A | |
A synthetic peptide made to a portion of human ubiquitin protein ligase E3A encoded within exon 6 (LocusLink ID 7337). | |
0.1 mL | |
Apoptosis, Transcription Factors and Regulators | |
7337 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction