Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E74 like factor 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP310893100UL

 View more versions of this product

Catalog No. NB126687


Only null left
Add to Cart

Description

Description

E74 like factor 1 Polyclonal specifically detects E74 like factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

E74 like factor 1
Polyclonal
Western Blot 1.0 ug/ml, Immunohistochemistry
E74-like factor 1, E74-like factor 1 (ets domain transcription factor), ETS-related transcription factor Elf-1
The immunogen is a synthetic peptide directed towards the N terminal region of human E74 like factor 1 (NP_758961). Peptide sequence VTLDGIPEVMETQQVQEKYADSPGASSPEQPKRKKGRKTKPPRPDSPATT
100 μg
Cellular Markers
1997
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.