Learn More
Invitrogen™ EAAC1 Monoclonal Antibody (N180/41), FITC
Description
5 μg of MA5-45548 was sufficient for immunoprecipitation of EAAC1 from 50 μg of rat brain lysate. Detects approximately 60kDa. This antibody was formerly sold as clone S180-41.
Specifications
Specifications
| Antigen | EAAC1 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N180/41 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 |
| Gene | Slc1a1 |
| Gene Accession No. | P43005, P51907 |
| Gene Alias | D130048G10Rik; DCBXA; EAAC1; EAAC2; EAAT3; excitatory amino acid carrier 1; excitatory amino acid carrier 2; excitatory amino acid transporter 3; excitatory amino acid transporter-3; excitatory amino-acid carrier 1; glutamate transporter mEAAC2; MEAAC1; Neuronal and epithelial glutamate transporter; REAAC1; SCZD18; SLC1A1; Sodium-dependent glutamate/aspartate transporter 3; solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1; Solute carrier family 1 A1 (brain glutamate transporter); solute carrier family 1 member 1; solute carrier family 1, member 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.