Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAC1 Polyclonal Antibody
EAAC1 Polyclonal Antibody
Supplier: Thermo Scientific PA595144
Description
EAAC1 Polyclonal Antibody for Western Blot

Specifications
EAAC1 | |
Polyclonal | |
Unconjugated | |
Slc1a1 | |
D130048G10Rik; DCBXA; EAAC1; EAAC2; EAAT3; excitatory amino acid carrier 1; excitatory amino acid carrier 2; excitatory amino acid transporter 3; excitatory amino acid transporter-3; excitatory amino-acid carrier 1; glutamate transporter mEAAC2; MEAAC1; Neuronal and epithelial glutamate transporter; REAAC1; SCZD18; SLC1A1; Sodium-dependent glutamate/aspartate transporter 3; solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1; Solute carrier family 1 A1 (brain glutamate transporter); solute carrier family 1 member 1; solute carrier family 1, member 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6505 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P43005 | |
Slc1a1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human EAAT3 (10-27aa EWKRFLKNNWVLLSTVAA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction