Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305771MFV450
Description
EAAT1/GLAST-1/SLC1A3 Polyclonal antibody specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
EAAT1/GLAST-1/SLC1A3 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
Partial recombinant protein made to an internal portion of the human SLC1A3 protein (between amino acids 100-300) [UniProt P43003] | |
0.1 mL | |
Cellular Markers, Cognition and Behavior | |
6507 | |
Store at 4C in the dark. |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction