Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298747MFV610
Description
EAAT1/GLAST-1/SLC1A3 Polyclonal antibody specifically detects EAAT1/GLAST-1/SLC1A3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EAAT1/GLAST-1/SLC1A3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the MOUSE EAAT1/GLAST-1/SLC1A3. | |
0.1 mL | |
Cellular Markers, Cognition and Behavior | |
6507 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction