Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody (SR2039), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321738100UL
Description
EAAT1/GLAST-1/SLC1A3 Monoclonal antibody specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
EAAT1/GLAST-1/SLC1A3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2039 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
A synthesized peptide derived from human EAAT1/GLAST-1/SLC1A3 (Uniprot #: P43003) | |
100 μg | |
Cellular Markers, Cognition and Behavior | |
6507 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction