Learn More
Eaat1 Polyclonal Antibody, Rockland™

Chicken Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Eaat1 |
---|---|
Concentration | 0.99 mg/mL |
Applications | ELISA, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1308
![]() ![]() |
Rockland Immunochemicals
200901ML1 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1309
![]() ![]() |
Rockland Immunochemicals
200901ML1S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against rat Slc1a3. This product is an IgY fraction antibody purified from monospecific chicken egg yolks by a multi-step process which includes selective precipitation and salt fractionation followed by extensive dialysis against the buffer stated above. Blast analysis the immunogen sequence shows 100% identity with mouse and 91.7% human.
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.Specifications
Eaat1 | |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Chicken | |
Rat | |
P24942 | |
29483 | |
Anti-Slc1a3 antibody was prepared from eggs of chickens laid after repeated immunizations with a synthetic peptide corresponding to a N-Terminal portion of rat Slc1a3 conjugated to Keyhole Limpet Hemocyanin (KLH). | |
-20°C, Avoid Freeze/Thaw Cycles | |
SLC1A3 |
0.99 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.002M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7 | |
AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; Glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; Solute carrier family 1 member 3; solute carrier family 1, member 3 | |
SLC1A3 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.