Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT1 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502206829
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6829 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6829 Supplier Fabgennix Inc Supplier No. EAAT1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EAAT1 Polyclonal antibody specifically detects EAAT1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen EAAT1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SLC1A3
Gene Accession No. P24942, P43003, P56564
Gene Alias AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; solute carrier family 1 member 3; solute carrier family 1, member 3
Host Species Rabbit
Immunogen Synthetic peptide taken wtihin amino acid region 475-550 on mouse EAAT1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20512, 29483, 461918, 6507
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.