Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT2/GLT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP159632
Description
EAAT2/GLT1 Polyclonal specifically detects EAAT2/GLT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EAAT2/GLT1 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml | |
P43004 | |
SLC1A2 | |
Synthetic peptides corresponding to SLC1A2(solute carrier family 1 (glial high affinity glutamate transporter), member 2) The peptide sequence was selected from the N terminal of SLC1A2 (NP_004162). PGNPKLKKQLGPGKKNDEVSSLDAFLDLIRNLFPENLVQACFQQIQTVTK The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
Rabbit | |
62 kDa | |
100 μL | |
Hypoxia, Neuroscience, Neurotransmission | |
6506 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction