Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT2/GLT1 Antibody (SR2082), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | EAAT2/GLT1 |
---|---|
Clone | SR2082 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
EAAT2/GLT1 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
A synthesized peptide derived from human EAAT2/GLT1 (Uniprot #: P43004) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2082 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Hypoxia, Neuroscience, Neurotransmission | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6506 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title