Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eaat2 Polyclonal Antibody, Rockland™
SDP

Goat Polyclonal Antibody

$174.73 - $636.24

Specifications

Antigen Eaat2
Concentration 1.05 mg/mL
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-1593
SDP
Encompass_Preferred
View Documents
Rockland Immunochemicals
600101MN3
100 μg
Each for $636.24
Only null left
Add to Cart
 
50-199-1594
SDP
Encompass_Preferred
View Documents
Rockland Immunochemicals
600101MN3S
25 μL
Each for $174.73
Only null left
Add to Cart
 
Description

Description

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against rat Eaat2. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to human, mouse, and rat.

Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.
Specifications

Specifications

Eaat2
ELISA, Immunofluorescence, Western Blot
Unconjugated
Goat
Mouse, Rat
P31596, P43006
20511, 29482
Primary
Antibody
1.05 mg/mL
Polyclonal
Liquid
RUO
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
1700091C19Rik; 2900019G14Rik; AI159670; EAAT2; EAAT2 variant mEAAT2/5UT5; Excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; Glt; GLT1; GLT-1; GLT1a; GLT1b; GLT1c; glutamate transporter 1c; Glutamate/aspartate transporter II; GLUT-R; HBGT; high affinity glucose transporter; MGLT1; OTTHUMP00000165209; OTTHUMP00000165210; SLC1A2; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; Solute carrier family 1 member 2; solute carrier family 1, member 2
SLC1A2
-20°C, Avoid Freeze/Thaw Cycles
SLC1A2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.