Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT3 Rabbit anti-Human, Mouse, Rat, Clone: JU39-69, Novus Biologicals™
SDP

Catalog No. NBP275476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275476 Supplier Novus Biologicals Supplier No. NBP275476

Rabbit Monoclonal Antibody

EAAT3 Monoclonal specifically detects EAAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EAAT3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JU39-69
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Gene Symbols SLC1A1
Host Species Rabbit
Immunogen Recombinant protein within Human EAAT3 aa 80-209/ 524. (SwissProt: P43005 Human; SwissProt: P51906 Mouse; SwissProt: P51907 Rat)
Molecular Weight of Antigen 57 kDa
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6505.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.