Learn More
Invitrogen™ EAF1 Monoclonal Antibody (1G2)

Description
Peptide Sequence: MNGTANPLLD REEHCLRLGE SFEKRPRASF HTIRYDFKPA SIDTSCEGEL QVGKGDEVTI TLPHIPGSTP PMTVFKGNKR PYQKDCVLII NHDTGEFVLE KLSSSIQVKK TRAEGSSKIQ ARMEQQPTRP PQTSQPPPPP PPMPFRAPTK PPVGPKTSPL KDNPSPEPQL DDIKRELRAE VDIIEQMSSS SGSSSSDSES SSGSDDDSSS SGGEDNGPAS PPQPSHQQPY NSRPAVANGT SRPQGSNQLM NALRNDLQLS ESGSDSDD.
Specifications
Specifications
| Antigen | EAF1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EAF1 |
| Gene Accession No. | Q96JC9 |
| Gene Alias | 4933403C17Rik; AI666536; EAF1; ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL associated factor 1; ELL-associated factor 1 |
| Gene Symbols | EAF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.