Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180575
Description
EAF2 Polyclonal specifically detects EAF2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EAF2 | |
Polyclonal | |
Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
NP_060926.2 | |
EAF2 | |
The immunogen this antibody was made to, maps to a region between residue 150 and 200 of human ELL associated factor 2 using the numbering given in entry NP_060926.2 (GeneID 55840). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
BM040, ELL associated factor 2, testosterone regulated apoptosis inducer and tumor suppressor, Testosterone-regulated apoptosis inducer and tumor suppressor protein, TRAITSELL-associated factor 2, U19 | |
Rabbit | |
Affinity Purified | |
RUO | |
55840 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction