Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113302
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113302 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113302 Supplier Invitrogen™ Supplier No. PA5113302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EAF2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

EAF1 and EAF2 can form a complex with ELL that targets the ternary elongation complex of Pol II, stimulating the rate of elongation. In addition, EAF1 and EAF2 are important for the stability of the NuA4 histone acetyltransferase complex, which transcriptionally activates certain genes by acetylation of Histones H4 and H2A. Both EAF1 and EAF2 are ubiquitously expressed members of the EAF family that colocalize with ELL to the the Cajal bodies and nuclear speckles. EAF1 contains a C-terminal region rich in aspartic acid, glutamic acid and serine residues. EAF2 is an androgen-response gene and can act as a potent apoptosis inducer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Eaf2
Gene Accession No. Q91ZD6, Q96CJ1
Gene Alias AW048865; BM040; BM-040; Eaf2; Ehrlich S-II transcriptional activator factor; ELL associated factor 2; ELL-associated factor 2; Festa; FESTA-L; FESTA-S; testosterone regulated apoptosis inducer and tumor suppressor; testosterone-regulated apoptosis inducer and tumor suppressor protein; Traits; U19
Gene Symbols Eaf2
Host Species Rabbit
Immunogen Recombinant Human ELL-associated factor 2 protein (6-100AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106389, 55840
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.