Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eaf6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30331920UL
Description
Eaf6 Polyclonal specifically detects Eaf6 in Human, Mouse samples. It is validated for Western Blot.Specifications
Eaf6 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
64769 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
CENP-28, centromere protein 28, chromatin modification-related protein MEAF6, MYST/Esa1-associated factor 6, NY-SAR-91, sarcoma antigen NY-SAR-91 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-191 of human Eaf6 (NP_001257804.1). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction